Smart Gasket Sanitary Sampling Gasket, Viton, 1 Port, 3/4" Tri-Clamp
Representative image only.
Smart Gasket Mfr # G-TH-075-V-1 – Item # WZ-30542-06
Smart Gasket lets you take samples or readings from a sanitary line
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
PRODUCT OPTIONS
£48.76GBP / Each
£58.51GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialViton
  • Connection TypeSanitary
  • Number Of Ports1
  • Sanitary Clamp Size (Port 1) (in)3/4
  • Description3/4" Tri-Clamp, 1 Port, Viton, Each
Key Features
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
More About this Item
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.