Sludge Judge Ultra C09251WA Replacement Middle/Extension Section
Representative image only.
Sludge Judge Mfr # B01388 – Item # WZ-05465-42
Keep your samplers running smoothly
  • Sludge Judge Ultra Series
  • 1 year warranty
PRODUCT OPTIONS
£88.32GBP / Each
£105.98GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Material3/4\" ID Polycarbonate tube
  • Sample Volume3 oz. per foot or 15 oz.
  • Length (in)60
  • Length (ft)5
  • Length (cm)152.4
  • DescriptionReplacement Middle/Extension Section
  • Warranty1 year
1 Year Warranty
Key Features
  • Sludge Judge Ultra Series
  • 1 year warranty
More About this Item
The following parts and accessories will help you keep your Sludge Judge samplers working properly.