Simpson 1329AT 04459 Taut Baud Dc Microammeter 0-10 Model:04459
Representative image only.
Simpson Mfr # 04459 – Item # WZ-68489-81
£246.14GBP / Each
£295.37GBP (incl VAT)
£107.32 - £301.63GBP / Each
£128.78 - £361.96GBP (incl VAT)
for an additional
£115.57GBP / Each
£138.68GBP (incl VAT)
for an additional
£107.32GBP / Each
£128.78GBP (incl VAT)
for an additional
£242.89GBP / Each
£291.47GBP (incl VAT)
for an additional
£149.19GBP / Each
£179.03GBP (incl VAT)
for an additional
£165.86GBP / Each
£199.03GBP (incl VAT)
for an additional
£301.63GBP / Each
£361.96GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Display Type4.5\" analog dial
  • Max DC Current mA (mA)0.01
  • Min DC Current mA (mA)0
  • DescriptionTaut Baud Dc Microammeter 0-10 Model:04459
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis