This item is now discontinued. Please browse similar items

Simpson 1329A 06540 Dc Milliammeter 0-50 4 1/2in Model:06540

Representative image only.
Simpson Mfr # 1329A | 06540 – Item # WZ-68488-80

SPECS & DESCRIPTION

Specifications & Description

  • Display Type4.5\" analog dial
  • Max DC Current mA (mA)50
  • Min DC Current mA (mA)0
  • DescriptionDc Milliammeter 0-50 4 1/2in Model:06540

More About this Item

FEATURES

  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized

Additional ranges available. Contact Davis

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat