Simpson 1329 04540 Dc Microammeter 100-0-100 4 1/2in Model:04540
Representative image only.
Simpson Mfr # 04540 – Item # WZ-68488-58
£137.28GBP / Each
£164.74GBP (incl VAT)
£107.32 - £301.63GBP / Each
£128.78 - £361.96GBP (incl VAT)
for an additional
£115.57GBP / Each
£138.68GBP (incl VAT)
for an additional
£107.32GBP / Each
£128.78GBP (incl VAT)
for an additional
£242.89GBP / Each
£291.47GBP (incl VAT)
for an additional
£149.19GBP / Each
£179.03GBP (incl VAT)
for an additional
£165.86GBP / Each
£199.03GBP (incl VAT)
for an additional
£301.63GBP / Each
£361.96GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Display Type4.5\" analog dial
  • Max DC Current mA (mA)10000000
  • Min DC Current mA (mA)0
  • DescriptionDc Microammeter 100-0-100 4 1/2in Model:04540
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis