This item is now discontinued. We recommend the following as suggested replacements: WZ-15957-65
SHOWA 7005PFM Laboratory Grade Gloves, powder-free, medium
Representative image only.
SHOWA Mfr # 7005PFM – Item # WZ-86287-81
Reduce hand fatigue by keeping your hands cool and dry
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)4
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorLight Blue
  • ComplianceFDA 21 CFR 170-199
  • SterileNo
  • PowderedNo
  • DescriptionLaboratory Grade Gloves, powder-free, medium
Key Features
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
More About this Item
N-DEX® gloves are made from a special low-modulus, nonlatex, soft nitrile formulation. They offer greater tactile-sensitivity, comfort, dexterity, and puncture-resistance than latex gloves. All N-DEX® nitrile gloves are ambidextrous and blue in color.
General and Laboratory Grade N-DEX® Gloves are ideal for use anywhere you need to protect your hands from the environment. Box of 100.