This item is now discontinued. Please browse similar items
Setra SEC1-C-B3B-03R-00 Calibrator Module C for Transducer
Representative image only.
Setra Mfr # SEC1-C-B3B-03R-00 – Item # WZ-98075-95
Optimizer performance of your pressure transducer
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCalibrator Module C for Transducer
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
These accessories are compatible with the Setra 98075-XX series AccuSense™ High-Accuracy Pressure Transducers.
The SecureCal™ calibrator 98075-95 is used to calibrate transducers in the field—enables zero-tare and snap-span adjustment, factory calibration restore function, and two-line LCD/LED feedback.