Seepex W05NA1F0BD3034 Progressing Cavity Pump, Iron/Nitrile, STD, 45 psi, 46 GPM; AC
Representative image only.
Seepex Mfr # W05NA1F0WG3034 – Item # WZ-76803-34
Excellent chemical resistance, pulseless flow, and low shear
  • Easily move a variety of polymers and chemicals, and extremely adaptable to a wide range of industries
  • Gentle low-shear fluid transfer capable of pumping small particles without damaging product or pump
PRODUCT OPTIONS
£2,833.95GBP / Each
£3,400.74GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (GPM)46
  • Max Flow Rate (LPM)174.2
  • Min Flow Rate (LPM)0
  • Wetted MaterialsIron, Nitrile
  • DescriptionProgressing Cavity Pump, Iron/Nitrile, STD, 45 psi, 46 GPM; AC
Key Features
  • Easily move a variety of polymers and chemicals, and extremely adaptable to a wide range of industries
  • Gentle low-shear fluid transfer capable of pumping small particles without damaging product or pump
More About this Item
These pumps are of two main configurations: iron bodies with NBR stators and stainless steel (SS) bodies with Viton® stators. The two material combinations are ideal for many different industries including food & beverage, environmental, chemical & biochemical, pulp & paper, oil, gas & petrochemicals, agriculture, construction, biogas, ceramics, pharmaceutical and cosmetics . . . just to name a few!
The unique design of the rotor/stator combination provides many benefits—the smaller rotor cross section results in reduced thrust loads, the universal joint design offers improved NPSHR and low component wear, and the plug-in shaft connection allows for quick part change out. Listed below are the three most commonly asked-for drive configurations. The first is a standard AC motor (STD), the
second is a standard AC motor with an integrated controller to facilitate your changing requirements (IVD), and the third is the Spaggiari drive system (SPG).