Scienceware Transpette 8-Channel Transfer Pipettors, LDPE, Non-Sterile; 25/Pk
Representative image only.
Scienceware Mfr # F378730000 – Item # WZ-07897-06
Economical, single-use, 8-channel transfer pipettor
  • Ideal for use with samples in micro-wellplates
  • Clear plastic construction
  • 600 µl capacity per channel
  • 3 month warranty
£70.79GBP / Pkg of 25
£84.95GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)600
  • Max Volume (mL)0.6
  • Max Dispensing (mL)0.6
  • Overall Length (in)7.48
  • Overall Length (mm)190
  • Number Of Channels8
  • Body MaterialLDPE
  • Description8-Channel Transfer Pipettors, LDPE, Non-Sterile; 25/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Ideal for use with samples in micro-wellplates
  • Clear plastic construction
  • 600 µl capacity per channel
  • 3 month warranty
More About this Item
Use these multichannel transfer pipetters in place of expensive pipettors and pipette tips. Use to perform plate washing, media changes or supernatant collection. Dispose after "hot" uses to avoid radiation and contamination of other costly pipettors. Not autoclavable.