Scienceware H36904-2006 Sterileware Bent Handle Scoop, 4 oz, 10 pack

Representative image only.
Scienceware Mfr # H36904-2006 – Item # WZ-06263-53
  • 3 month warranty

PRODUCT OPTIONS

£41.92GBP / Pkg of 10
£50.30GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (oz)4.226
  • Capacity (mL)125
  • Width (in)5
  • Width (cm)12.7
  • MaterialPolystyrene
  • SterileYes
  • DescriptionSterileware Bent Handle Scoop, 4 oz, 10 pack
  • Warranty3 month
3 Month Warranty

Key Features

  • 3 month warranty

More About this Item

The curved handle of these scoops elevates the users hand to reduce the possibility of source contamination and exposure to hazardous substances. Made from durable polystyrene they are safe for food, drug, and cosmetic contact. A flat bottom helps keep the scoops upright allowing them to be used as a weight boat. Each individually packaged single use scoop is sterilized by gamma irradiation, lot stamped for accurate tracing, and shipped with certificate of sterilization.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat