This item is now discontinued. Please browse similar items
Scienceware F371400015 Spinring® Stir Bar, PTFE, 1-3/4" OD; 1/Ea
Representative image only.
Scienceware Mfr # F371400015 – Item # WZ-04775-44
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)5/16
  • Length (cm)0.8
  • Diameter (in)1.75
  • Diameter (cm)4.45
  • DescriptionSpinring® Stir Bar, PTFE, 1-3/4\" OD; 1/Ea
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Removable stabilizing ring keeps stir bar centered in vessel. Octagon shape creates intense vortex without splashing or chattering and resists spinout.