Scienceware F371250038 Spinfin Stirring Bar, PTFE, 3/8" X 3/8"
Representative image only.
Scienceware Mfr # F371250038 – Item # WZ-04775-33
  • 1 year warranty
PRODUCT OPTIONS
£10.73GBP / Each
£12.88GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.375
  • Diameter (cm)0.9525
  • DescriptionSpinfin Stirring Bar, PTFE, 3/8\" X 3/8\"
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Vigorously shakes liquids in round-bottom test tubes and flasks, where other stir bars cannot be used. Position is maintained in a vessel and will not vibrate.