Scienceware F371250034 Spinfin Stirring Bar, PTFE, 3/4" X 1/2"
Representative image only.
Scienceware Mfr # F371250034 – Item # WZ-04775-31
£15.99GBP / Each
£19.19GBP (incl VAT)
Cole-Parmer Clean-Wipes, 70% Alcohol/30% DI Water; 100/Canister
£8.50GBP / Pkg of 100
£10.20GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.5
  • Diameter (cm)1.27
  • DescriptionSpinfin Stirring Bar, PTFE, 3/4" X 1/2"
More About this Item
Vigorously shakes liquids in round-bottom test tubes and flasks, where other stir bars cannot be used. Position is maintained in a vessel and will not vibrate.