Scienceware F371250012 Spinfin Stirring Bar, PTFE, 1/2" X 1/2"
Representative image only.
Scienceware Mfr # F371250012 – Item # WZ-04775-27
  • 1 year warranty
PRODUCT OPTIONS
£11.67GBP / Each
£14.00GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)1.9685
  • Diameter (cm)5
  • DescriptionSpinfin Stirring Bar, PTFE, 1/2\" X 1/2\"
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Vigorously shakes liquids in round-bottom test tubes and flasks, where other stir bars cannot be used. Position is maintained in a vessel and will not vibrate.