This item is now discontinued. Please browse similar items
Scienceware F24880-4002 Plum Double Sterile Saline Eyewash Station, two 500 mL bottles
Representative image only.
Scienceware Mfr # F24880-4002 – Item # WZ-06796-33
Flexible, form-fitting eye cup ensures gentle and effective rinsing
  • Bendable eye cup keeps eyelid open during use
  • All solutions meet U.S. Federal Regulations, comply with ANSI standards, and are FDA compliant for personal eyewash
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPlum Double Sterile Saline Eyewash Station, two 500 mL bottles
Key Features
  • Bendable eye cup keeps eyelid open during use
  • All solutions meet U.S. Federal Regulations, comply with ANSI standards, and are FDA compliant for personal eyewash
More About this Item
Ideal for emergency rinsing of debris, and neutralization of acids or alkalis in the eyes. Bottles with a green eye cup are sterile saline eye wash; use to wash particulate debris from the eye. Bottles with blue eye cups contain a sterile phosphate buffer solution; use to rapidly neutralize acids, alkalis, and other chemicals in the eye.
To use simply twist the cap to open, apply the eye cup to the affected eye, and squeeze the bottle. Control the flow of solution with hand pressure on the bottle for more efficient rinsing and less waste. Each bottle of DUO eyewash releases two streams to rinse both eyes at once.
Compliance: ANSI Z358.1-2004