Sartorius Cubis II Analytical Balance, 320 g x 0.1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz
Representative image only.
Sartorius Mfr # MCE324SU-S00 – Item # WZ-39742-31
Individual configuration of draft shield and integrated ionizer provides adaptation to any weighing task
  • Monolithical weighing system, integrated climate sensors, individual sample holders offer industry leading performance
  • Individual QApp workflows and motorized auto-leveling reduce error
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management provides excellent data integrity
  • Innovative TFT touch screen operation
  • Factory installed set of essential weighing applications
  • Activated by touch keypad or touch-free using IR sensor (items 39742-30 and 39742-32)
  • Essential user interface (MCE) features high-contrast touch display for easy operation
  • 10 year warranty
PRODUCT OPTIONS
£9,194.29GBP / Each
£11,033.15GBP (incl VAT)
£194.13 - £201.02GBP / Each
£232.96 - £241.22GBP (incl VAT)
for an additional
£194.13GBP / Each
£232.96GBP (incl VAT)
for an additional
£201.02GBP / Each
£241.22GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Optimize balance performance with…
Optimize balance performance with…
Sartorius Software
£570.80 - £2,579.51GBP / Each
£684.96 - £3,095.41GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (g)320
  • Readability (mg)0.1
  • Repeatability (mg)0.04
  • Linearity (mg)0.2
  • Calibration TypeExternal
  • OutputRS-232, USB-A, USB-B
  • Pan Length (in)3 11/32
  • Pan Width (in)3 11/32
  • Draft Shield Height (in)10 15/64
  • Draft Shield Width (in)8 5/64
  • Display Height (in)14 11/16
  • Display Width (in)9 29/64
  • Draft Shield Depth (in)8 7/64
  • DescriptionAnalytical Balance, 320 g x 0.1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz
  • Warranty10 Years
10 Year WarrantyConformite EuropeenneCanadian Standards Association
Includes
Device, weighing pan, shield plate, power supply unit, country-specific power cable, USB cable, in-use dust cover for control unit, dust cover for analytical draft shield.
Key Features
  • Monolithical weighing system, integrated climate sensors, individual sample holders offer industry leading performance
  • Individual QApp workflows and motorized auto-leveling reduce error
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management provides excellent data integrity
  • Innovative TFT touch screen operation
  • Factory installed set of essential weighing applications
  • Activated by touch keypad or touch-free using IR sensor (items 39742-30 and 39742-32)
  • Essential user interface (MCE) features high-contrast touch display for easy operation
  • 10 year warranty
More About this Item
Analytical balances have a maximum capacity between 120 g and 520 g. The automatic glass draft shield with integrated ionizer removes interfering electrostatic charges on samples and sample containers. The manual glass draft shield has smooth-action doors. Large, high-contrast touch display with factory-installed essential weighing applications for easy operation.