Cole-Parmer Essentials 96-Place Reversible Tube Rack no Lid, PP, Assorted Colors; 5/PK

Representative image only.
Cole-Parmer Essentials Item # WZ-06736-09
Withstand temperatures down to -80 °C (-112 °F)
  • Autoclavable
  • 1 year warranty

PRODUCT OPTIONS

£37.23GBP / Pkg of 5
£44.68GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Tubes Held96
  • Length (in)8 3/8
  • Width (in)4 1/2
  • Height (in)1 1/16
  • MaterialPolypropylene(PP)
  • ColorAssorted
  • DescriptionReversible 96-well microcentrifuge tube rack, assorted colors
  • Warranty1 year
3 Month Warranty

Key Features

  • Autoclavable
  • 1 year warranty

More About this Item

One side of this polypropylene (PP) racks is designed to hold 0.5mL microcentrifuge tubes; the other side holds 1.5 to 2.0mL tubes. Withstand temperatures down to -80°C (-112°F).

Please note: lid not included.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat