This item is now discontinued. Please browse similar items
Red Lion G3CF256M CompactFlash Card, 256 MB
Representative image only.
Red Lion Mfr # G3CF256M – Item # WZ-68515-54
Human machine interfaces that strike the perfect balance of advanced features and value!
  • Unit's configuration is stored in nonvolatile Flash memory
  • NEMA 4/IP65 front panel
  • 3 front panel LED indicators
  • Resistive analog touch-screen
  • Bright TFT display with full 256-color support
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCompactFlash Card, 256 MB
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Unit's configuration is stored in nonvolatile Flash memory
  • NEMA 4/IP65 front panel
  • 3 front panel LED indicators
  • Resistive analog touch-screen
  • Bright TFT display with full 256-color support
  • 1 year warranty
More About this Item
Model G304K - Kadet Operator Interface with 4.3'' TFT Color Display
The 4.3-inch G304K was designed for applications in which available mounting space is at a premium. The G304K offers two high-speed serial ports in the form of one RS-232 and one RS-232/422/485 ports. This allows it to simultaneously communicate with devices from different manufacturers, as well as to perform protocol conversion.
Models G306K/G308K with 5.6''/8'' TFT Color Display
The G306K & G308K are the perfect solution for applications that require the operator to monitor and control more than just a single device. With three serial ports and an Ethernet port, either Kadet can connect to multiple serial and Ethernet devices simultaneously, including PLCs, motor drives, bar code scanners, etc. The G306K & G308K perform the functions of a multiple protocol converter, using three high-speed RS-232/422/485 communications ports and a 10 Base-T Ethernet port. The Ethernet port supports up to four protocols simultaneously, allowing dissimilar Ethernet-based products to communicate with one another.
All Kadet Operator Interfaces are programmed with Red Lion’s free Crimson 2.0 software.