This item is now discontinued. Please browse similar items
Red Lion G306A000 HMI 5.7" TFT Active Matrix QVGA 320x240 LCD, Indoor
Representative image only.
Red Lion Mfr # G306A000 – Item # WZ-68515-47
Benchmark of reliability and durability
  • Models come standard with one 10/100Base-T(X) Ethernet, two RS-232, and one RS-422/485 serial ports.
  • Communicates in over 300 different protocols.
  • Built-in data loggers for continuous data, event, and alarm collection and monitoring.
  • Data is compiled in CSV-formated files and can be stored, displayed, emailed, or automatically synchronized with a server.
  • A web server is built-in to allow remote control and monitoring.
  • Programming via Red Lion Crimson 3.0 software platform.
  • 32-bit ColdFire processor
  • Compact flash socket for data logging
  • NEMA 4X / IP66 front panel
  • 24 VDC operation
  • 8 MB non-volitile flash memory
  • 5.7" TFT Active Matrix 256 color QVGA 320 x 240 pixel LCD
  • Five button keypad for on-screen menus
  • Three front panel LED indicators
  • Resistive analog touchscreen
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHMI 5.7\" TFT Active Matrix QVGA 320x240 LCD, Indoor
  • Warranty1 Year
1 Year Warranty
Key Features
  • Models come standard with one 10/100Base-T(X) Ethernet, two RS-232, and one RS-422/485 serial ports.
  • Communicates in over 300 different protocols.
  • Built-in data loggers for continuous data, event, and alarm collection and monitoring.
  • Data is compiled in CSV-formated files and can be stored, displayed, emailed, or automatically synchronized with a server.
  • A web server is built-in to allow remote control and monitoring.
  • Programming via Red Lion Crimson 3.0 software platform.
  • 32-bit ColdFire processor
  • Compact flash socket for data logging
  • NEMA 4X / IP66 front panel
  • 24 VDC operation
  • 8 MB non-volitile flash memory
  • 5.7" TFT Active Matrix 256 color QVGA 320 x 240 pixel LCD
  • Five button keypad for on-screen menus
  • Three front panel LED indicators
  • Resistive analog touchscreen