This item is now discontinued. Please browse similar items
Red Lion BMK40000 BASE MOUNT KITAPOLLO/GEMINI
Representative image only.
Red Lion Mfr # BMK40000 – Item # WZ-68483-63
Provides mounting alternatives including DIN rail option
  • Compatible with CUB4, CUB4, or DT8 series meters
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBASE MOUNT KITAPOLLO/GEMINI
Key Features
  • Compatible with CUB4, CUB4, or DT8 series meters
More About this Item
These adapter kits are designed to offer an alternative mounting option for Red Lion’s CUB4 indicator/meters and CUB5 meters.
Easy to assemble and wire! The wires can either be brought through the panel on which the meter is mounted, or through the holes in the enclosure itself. Grommets are provided to insert in the holes, along with four nuts and bolts for mounting.
BMK6 open-base mount accommodates only the meter. BMK7 closed-base mounts are large enough to accommodate both the meter and a micro-line power supply (MLPS). Constructed of rugged steel with textured black finish.
BMK11 can be used to mount either a CUB5 meter or an MLPS in various applications — it provides both base mounting and DIN rail mounting. The adapter and DIN rail clips are constructed of molded polycarbonate plastic.