This item is now discontinued. Please browse similar items
Raven Environmental SID-20203-50 Replacement Probe for Wine Sediment Interface Detector; 50 Foot Cable
Representative image only.
Raven Environmental Mfr # SID-20203-50 – Item # WZ-05956-65
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With05956-60/-64
  • DescriptionReplacement Probe for Wine Sediment Interface Detector; 50 Foot Cable
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Replacement sludge and wine sediment interface detector sensors for your Raven Environmental samplers. Available with 20-, 30- and 50-ft cables.
The infrared sensor probe detects changes in density and sends the reading to the portable handheld meter. The probe has an integrated cable with bright-yellow depth markings for high visibility. Additionally, the stainless steel probe housing and polyurethane-coated cable are rugged, easy to clean, and made of USA FDA compliant materials.