This item is now discontinued. Please browse similar items
Raven Environmental SID-10203-20 Replacement Sludge Interface Detector Sensor, 20-ft cable
Representative image only.
Raven Environmental Mfr # SID-20203-20 – Item # WZ-05956-24
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With05956-20
  • DescriptionReplacement Sludge Interface Detector Sensor, 20-ft cable
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Replacement sludge and wine sediment interface detector sensors for your Raven Environmental samplers. Available with 20-, 30- and 50-ft cables.