Ralston Instruments QTHA-BLS0-HC Block and Bleed Valve
Representative image only.
Ralston Instruments Mfr # QTHA-BLS0-HC – Item # WZ-16107-14
Leak free connections for pressure calibrations
  • 1 year warranty
PRODUCT OPTIONS
£229.16GBP / Each
£274.99GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBlock and Bleed Valve
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High quality fitting kits and connection hoses are designed to give reliable and leak free connections for pressure calibrations. Since these fittings do not require wrenches or thread sealant, these fitting kits and quick connect hoses reduce calibration time and down time due to calibration of pressure gauges and transmitters.
The block and bleed valve is used to keep a calibration system pressurized with fluid or gas while venting pressure from the device being calibrated. One end is Quick-test (female) for connecting to the manifold while the other is a Quick-test (male). Stainless steel construction with Buna N seal.