Qsonica 419 Ultrasonic Cell Distruptor Microtip Probe 1/8" dia
Representative image only.
Qsonica Mfr # 4418 – Item # WZ-04711-90
  • 2 year warranty
PRODUCT OPTIONS
£378.00GBP / Each
£453.60GBP (incl VAT)
Cole-Parmer Threaded Ultrasonic Probe Tip, Stepped; 1/8
£360.93GBP / Each
£433.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Max Sample Size (mL)15
  • Tip Diameter (in)0.125
  • Tip Diameter (cm)0.3175
  • DescriptionUltrasonic Cell Distruptor Microtip Probe 1/8\" dia
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.