This item is now discontinued. Please browse similar items
QLA PEAK Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Amber Glass; Serialized
Representative image only.
Mfr # GLA9PK-A70XDS – Item # WZ-41306-49
QLA vessels provide more precise and consistent dissolution results
  • Unique serial number is laser marked on plastic rim of each vessel
  • Plastic rims help minimize glass breakage
  • 1000 mL capacity
  • PEAK-style vessel
  • Amber glass is ideal for light-sensitive compounds
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)1000
  • MaterialGlass
  • Bottle ColorAmber
  • Special FeaturesPEAK style with plastic rim
  • DescriptionPEAK Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Amber Glass; Serialized
Key Features
  • Unique serial number is laser marked on plastic rim of each vessel
  • Plastic rims help minimize glass breakage
  • 1000 mL capacity
  • PEAK-style vessel
  • Amber glass is ideal for light-sensitive compounds
More About this Item
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle.
Not available for sale in the United States.
PEAK vessels do not comply with USP specifications and cannot be substituted in existing applications.