QLA Hinged Cover Compatible with Agilent 708/709-DS Baths, Gray
Representative image only.
Mfr # COVERV-708 – Item # WZ-41308-30
Reduce loss of media due to evaporation
  • Protect samples from contamination or spillage
  • Specific styles and colors assist almost every process
  • For Agilent 708/709-DS baths
  • Gray
PRODUCT OPTIONS
£151.51GBP / Each
£181.81GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ColorGray
  • Compatible WithAgilent 708/709-DS baths
  • DescriptionHinged Cover Compatible with Agilent 708/709-DS Baths, Gray
Key Features
  • Protect samples from contamination or spillage
  • Specific styles and colors assist almost every process
  • For Agilent 708/709-DS baths
  • Gray
More About this Item
Vessel covers reduce the potentially considerable loss of media due to evaporation. Standard and low-loss vessel covers are available for most dissolution baths. Cover styles include all common one-piece and hinged covers, as well as a unique rotary configuration. Industry standard openings are provided to accommodate cannula and stoppers.
Not available for sale in the United States.