Pyrex Vista 70024-50 Graduated Glass Cylinder, 50 mL, to deliver, 1/ea
Representative image only.
Pyrex Vista Mfr # 70024-50 – Item # WZ-34504-72
Calibrated "to deliver"
PRODUCT OPTIONS
£31.85GBP / Each
£38.22GBP (incl VAT)
£90.44 - £142.45GBP / Each
£108.53 - £170.94GBP (incl VAT)
for an additional
£90.44GBP / Each
£108.53GBP (incl VAT)
for an additional
£142.45GBP / Each
£170.94GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)50
  • Graduation Interval (mL)1
  • Tolerance (mL)±0.50
  • DescriptionGraduated Glass Cylinder, 50 mL, to deliver, 1/ea
More About this Item
Manufactured of Class A borosilicate glass, these metric scale cylinders are enameled with blue graduations for easy reading. Cylinder capacity is in accordance wtih ASTM E-1272. 10mL size has a funnel top for easy filling.