This item is now discontinued. Please browse similar items
Pyrex 3920-M Liquid Extractor for Fluids Heavier than Water, Glass, 1 L
Representative image only.
Pyrex Mfr # 3920-M – Item # WZ-34510-10
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLiquid Extractor for Fluids Heavier than Water, Glass, 1 L
More About this Item
Extractor for fluids heavier than water is used with solvents of heavier specific gravity than sample liquid being extracted. Includes extractor, Allihn condenser and boiling flask. Meets EPA, RCRA, and CLP standards. Total height: 710 mm.