This item is now discontinued. Please browse similar items
Puma AT-7018 Air Orbital Sander, 3-2/3 in x 6-3/4 in pad, 8,000 rpm, 1/4 in connection, 90 psi
Representative image only.
Puma Mfr # AT-7018 – Item # WZ-07122-18
  • Different pad shapes and sizes suitable for a wide range of applications
  • Ideal for sanding, polishing and buffing
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Free Air Capacity (CFM)6
  • Max Pressure (PSI)90
  • DescriptionAir Orbital Sander, 3-2/3 in x 6-3/4 in pad, 8,000 rpm, 1/4 in connection, 90 psi
  • Warranty1 year
1 Year Warranty
Key Features
  • Different pad shapes and sizes suitable for a wide range of applications
  • Ideal for sanding, polishing and buffing
  • 1 year warranty
More About this Item
Make the most of your air compressor with these air-powered tools. All require 90 psi pressure, but vary in air consumption.