Dynalon PTFE Joint Sleeves for Standard Taper Size 24/40; 6/Pk
Representative image only.
Dynalon Mfr # 362635 – Item # WZ-06073-40
  • 1 year warranty
PRODUCT OPTIONS
£61.10GBP / Pkg of 6
£73.32GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Taper Size24/40
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionPTFE Joint Sleeves for Standard Taper Size 24/40; 6/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Translucent PTFE, self-lubricating sleeves are precision tapered for a snug fit. Withstand temperatures from –103 to 482°>F (–75 to 250°>C). Pack of 6. 3 pk/cs*.