This item is now discontinued. Please browse similar items
Properties of Gases and Liquids, Fifth Edition
Representative image only.
Mfr # 0-07-011682-2 PT – Item # WZ-00541-04
SPECS & DESCRIPTION
Specifications & Description
  • ISBN0-07-011682-2
  • AuthorB. Poling, J. Prausnitz, and J. O'Connell
  • Edition5
  • Published Date2000
  • Number of Pages768
  • PublisherMcGraw-Hill Professional
  • DescriptionProperties of Gases and Liquids, Fifth Edition
More About this Item
This book is a comprehensive and critical survey of the most reliable estimating methods in use today—plus property values for more than 600 pure chemicals. This book includes a critical analysis of existing estimating methods as well usable, practical recommendations.