This item is now discontinued. Please browse similar items
Process Technology VX.3211 Immersion Heater, Two PTFE Coated Rod s, 6" L Heated Area, 300 Watts, 240 V
Representative image only.
Process Technology Mfr # VX.3211 – Item # WZ-36136-15
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Immersion Depth (in)11
  • Heated Area6 in L
  • Max Temperature (° F)450
  • Max Temperature (° C)232
  • Min TemperatureAmbient;Ambient
  • Plug TypeNo plug
  • Power (VAC)240
  • Power (watts)300
  • DescriptionImmersion Heater, Two PTFE Coated Rod s, 6\" L Heated Area, 300 Watts, 240 V
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Multi-Element Heaters Encased in 304 Stainless Steel or PTFE include a grounded solid-state heating element. Heaters are self-limiting for safer operation. PTFE-coated heaters are ideal for use in corrosive environments. The numbers of heating rods coming down from the polypropylene terminal head are dependent on the wattage of the heater: 300 Watt = 2 rods, 500 Watt = 3 rods, and 1000 Watt = 5 rods. Heaters include 3 ft wire and flexible conduit.