This item is now discontinued. Please browse similar items
Process Technology VS.1207 Immersion Heater, 304 SS Bayonet, 4" L Heated Area, 100 Watts, 240 V
Representative image only.
Process Technology Mfr # VS.1207 – Item # WZ-36135-05
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Immersion Depth (in)7
  • Heated Area4 in L
  • Max Temperature (° F)450
  • Max Temperature (° C)232
  • Min TemperatureAmbient;Ambient
  • Plug TypeNo plug
  • Power (VAC)240
  • Power (watts)100
  • DescriptionImmersion Heater, 304 SS Bayonet, 4\" L Heated Area, 100 Watts, 240 V
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Single Element Heaters Encsed in 304 Stainless Steel or PTFE include a grounded solid-state heating element. Heaters are self-limiting for safer operation. Simple connection and compact design make these heaters ideal—even where space is limited. Heaers include 4-ft power cord with a three-prong plug; 240 VAC models do not include a plug.