PRO Scientific Multi-Gen 7XL Stainess Steel Probes; 12/Pk
Representative image only.
PRO Scientific Mfr # 02-070MGXL-12 – Item # WZ-04751-70
Expand and upgrade your homogenizing capabilities
  • 2 year warranty
PRODUCT OPTIONS
£1,919.55GBP / Pkg of 12
£2,303.46GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)15
  • Material316 Stainless Steel & PTFE
  • Compatible WithPRO Scientific Homogenizers
  • Length (mm)115
  • Diameter (mm)7
  • DescriptionStainess Steel Probes; 12/Pk
  • Warranty2 year
Two year warrantyMade in the U.S.A.
Key Features
  • 2 year warranty
More About this Item
Parts and accessories compatible with PRO Scientific homogenizers.
Multi-Gen 7XL Generator Probe System combines an adapter (04751-69) and a pack of twelve 316 stainless steel and PTFE generators (04751-70) to replace disposable plastic generators. Adapter is compatible with most PRO Scientific homogenizer models including Bio-Gen PRO200, PRO250, PRO25D, SB-1, and PRO400DEL. The generator probes feature an open slotted tooth design suitable for processing even tough tissue samples up to 15 mL, and can be sterilized by autoclave or flame. Unlimited reusability make this system an economical choice when cross-contamination is a serious concern.