This item is now discontinued. Please browse similar items
PRO Scientific Deflector Head for Homogenizer Probe, 37 mm; Stainless Steel
Representative image only.
PRO Scientific Mfr # 02-37000S – Item # WZ-04751-89
Expand and upgrade your homogenizing capabilities
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • Compatible WithPRO Scientific Generator Probes, 37 mm diameter
  • Diameter (mm)37
  • DescriptionDeflector Head for Homogenizer Probe, 37 mm; Stainless Steel
  • Warranty2 year
Two year warrantyMade in the U.S.A.
Key Features
  • 2 year warranty
More About this Item
Parts and accessories compatible with PRO Scientific homogenizers.
Deflector Heads enhance homogenization by increasing sample turnover. Ideal for large or viscous samples, the design creates vertical movement of the sample from the generator tip, repeatedly cycling the material up and down. Choose between stainless steel and acetal resin construction.