This item is now discontinued. Please browse similar items

Precision Digital PD683-0K0 Loop-Powered Indicator, general purpose

Representative image only.
Precision Digital Mfr # PD683-0K0 – Item # WZ-61600-00
Designed for demanding process control applications
  • 5-digit backlit display with bargraph
  • Custom engineering units
  • HART® protocol transparent
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • DIN Size1/8\"
  • Control Input mA4-20
  • Display Type0.6\" backlit LCD
  • Display Digits5
  • Max Operating Temperature (° F)149
  • Min Operating Temperature (° F)-4
  • EnclosureHigh-impact plastic, uL 94V-0
  • Overall Width (in)4 43/64
  • Overall Height (in)2 29/64
  • Overall Depth (in)3 51/64
  • Overall Width (mm)118.9
  • Overall Height (mm)62.2
  • Overall Depth (mm)96.3
  • DescriptionLoop-Powered Indicator, general purpose
  • Warranty3 Years
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationFM ApprovedIntrinsically Safe

Key Features

  • 5-digit backlit display with bargraph
  • Custom engineering units
  • HART® protocol transparent
  • 3 year warranty

More About this Item

The Loop Leader Series’ PD683 & PD688 were designed for demanding process control applications. Standard features include a full 5-digit backlit display with bargraph, on-screen custom engineering units, and max/min display capabilities. Square root and programmable exponent functions are standard features on these meters. With these features these indicators can display flow from a DP transmitter or open channel flow in weirs and flumes. Programming is easy through the front panel buttons, with no pots or jumpers to deal with, and can be scaled without a signal source. The Loop Leader Series meters go just about anywhere. The PD688, with FM, CSA and ATEX certifications, can be installed in just about any hazardous area.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat