Precision Digital PD6801 Explosion-Proof Feet/Inches Level Meter

Representative image only.
Precision Digital Mfr # PD6801-0K1 – Item # WZ-30004-92
Displays level in feet and inches
  • 32-point linearization
  • Tank level bar graph
  • Lower display for custom tag, volume, or percent
  • Open collector output for high or low alarm

£719.00GBP / Each
£862.80GBP (incl VAT)

£96.00GBP / Each
£115.20GBP (incl VAT)

for an additional
£96.00GBP / Each
£115.20GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Control Input mA4-20
  • Display Type0.6" LED; 0.4" LED
  • Max Operating Temperature (° F)167
  • Min Operating Temperature (° F)-40
  • EnclosureNEMA 4X, 7, 9; IP68; explosion-proof die-cast aluminum
  • Overall Width (in)5
  • Overall Height (in)5 1/4
  • Overall Depth (in)4
  • Overall Width (mm)142.9
  • Overall Height (mm)133.4
  • Overall Depth (mm)123.8
  • DescriptionExplosion-Proof Feet/Inches Level Meter

Key Features

  • 32-point linearization
  • Tank level bar graph
  • Lower display for custom tag, volume, or percent
  • Open collector output for high or low alarm

More About this Item

The ProtEX-F&I PD6801 explosion-proof level meter brings enhanced functionality and readability to hazardous areas in a unique way — it displays level in feet and inches, not a decimal, so it can be read and understood effortlessly. The meter can display in eighths or sixteenths of an inch. Display features a backlight, and can be seen under varied lighting conditions, from wide angles, and from a distance. Program the meter using SafetTouch® thru-glass buttons, eliminating the need to remove the cover.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat