This item is now discontinued. Please browse similar items
Powerblanket PBICE15IP Insulating Cooling Blanket with Ice Packs; 15 Gallon
Representative image only.
Powerblanket Mfr # PBICE15IP – Item # WZ-42025-26
Innovative cooling blanket provides long lasting portable cooling
  • Easily control temperature with ice pack count
  • Advanced insulation directs heat away from vessel keeping temperature sensitive materials cool
  • No power required —portable cooling blankets can be used anywhere
  • Designed to accomidate 15 gallon bucket—blanket is 14.4" x 25"
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Drum Size15 gallons
  • Width (in)25
  • Length (in)14 2/5
  • DescriptionInsulating Cooling Blanket with Ice Packs; 15 Gallon
  • Warranty1 Year
1 Year Warranty
Includes
12 ice packs.
Key Features
  • Easily control temperature with ice pack count
  • Advanced insulation directs heat away from vessel keeping temperature sensitive materials cool
  • No power required —portable cooling blankets can be used anywhere
  • Designed to accomidate 15 gallon bucket—blanket is 14.4" x 25"
  • 1 year warranty
More About this Item
These insulating cooling drum and pail blankets are an easy and cost-effective way to cool large vessels containing temperature sensitive materials. Simply place the ice packs in the pockets on the inside surface of the blanket and place the blanket on to the drum. Modify the number of ice packs used to regulate temperature. Built-in straps securely cinch blanket to the outside of the drum for optimum performance and enhanced portability.