This item is now discontinued. Please browse similar items
Powerblanket Beacon TC-W3010 Smart Temperature Controller; 24 Amps
Representative image only.
Powerblanket Mfr # TCW3010 – Item # WZ-42025-54
Control and monitor temperatures remotely
  • Receive wireless alerts and notifications
  • Adjustable short cycle delay for refrigeration applications
  • Watertight—NEMA 4X, rugged, steel-enclosed casing
  • Backlit LCD makes for easy viewing
  • Maximum amperage of 24 amps
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (° F)212
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (DegreeC)100
  • Alarm OutputYes
  • Data LoggingYes
  • Enclosure RatingNEMA 4X
  • Display TypeLCD
  • Power (VAC)120
  • DescriptionSmart Temperature Controller; 24 Amps
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Includes
Two temperature probes with 9.8-ft (3 m) leads.
Key Features
  • Receive wireless alerts and notifications
  • Adjustable short cycle delay for refrigeration applications
  • Watertight—NEMA 4X, rugged, steel-enclosed casing
  • Backlit LCD makes for easy viewing
  • Maximum amperage of 24 amps
  • 1 year warranty
More About this Item
Compatible with a wide variety of heating or cooling devices, the Wi-Fi connected temperature controllers allow you to remotely monitor current conditions and adjust setpoints for your devices from a smartphone or computer using a web portal or mobile app. Receive wireless alerts and notifications when the temperature or other values go outside your defined thresholds. Alerts can be customized based on temperature, electrical current, voltage, and resistance readings. Easily log, access, and export historic temperature data for future reference or process analysis. Electrical current and voltage can also be collected, allowing you to analyze the duty cycle, energy usage, and operating cost of your cooling or heating equipment.
Local and remote access can be secured with user-specific PIN numbers. You can also assign multiple users login access with read-write or read-only permission levels.