This item is now discontinued. Please browse similar items
Portable Water Depth Sounder Gauge
Representative image only.
Item # WZ-99756-40
Instantly gauges the depth of a body of water
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Battery TypeOne 9 V (included)
  • Length (in)7 3/4
  • Width (in)1 3/4
  • DescriptionPortable Water Depth Sounder Gauge
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Gauge measures through air-free boat hulls, ice, or any solid surface in contact with water. Waterproof gauge will float if dropped. Updates reading every 10 seconds.