This item is now discontinued. Please browse similar items
Porta-Lab MF-2001 Portable Butane Microburner
Representative image only.
Porta-Lab Mfr # MF-2001 – Item # WZ-36310-47
Provides safe, portable flame applications in any work area
  • Burner requires no wicks to replace or gas lines to install
  • Refills quickly from a standard butane fuel cartridge
  • Stable tip-resistant design with easy-grip handle for safety
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeButane
  • Width (in)5
  • Height (in)3 1/2
  • DescriptionPortable Butane Microburner
  • Warranty1 Year
1 Year Warranty
Key Features
  • Burner requires no wicks to replace or gas lines to install
  • Refills quickly from a standard butane fuel cartridge
  • Stable tip-resistant design with easy-grip handle for safety
  • 1 year warranty
More About this Item
This portable microburner operates with a push-button auto ignition and features an adjustable flame size. Burner measures 5''W x 3-1⁄2''H. The optional tripod stand is made of high quality chrome on steel components to resist corrosion and discoloration, and supports a 1000-mL beaker.