This item is now discontinued. Please browse similar items
PolyScience MX27VB5G-A12 Constant-Temperature Viscosity Baths, 27 L, 240 VAC
Representative image only.
PolyScience Mfr # MX27VB6G-A12E – Item # WZ-98935-28
An economical alternative for kinematic viscosity measurement
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Reservoir Capacity (L)27
  • Max Temperature (° C)135
  • Min Temperature (° C)Ambient
  • Temperature SettingDigital
  • Power (VAC)230
  • DescriptionConstant-Temperature Viscosity Baths, 27 L, 240 VAC
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • 2 year warranty
More About this Item
These baths are ideal for measuring petroleum, lubricants, polymer coatings, food, paint, and other liquids' kinematic viscosity. Advanced PID controls maintain precise temperatures and quick responsiveness under changing heat load conditions. The integral recirculating pump ensures uniform temperature. Large, backlit digital display shows both set and actual values. Other features include Celsius/Fahrenheit selection, high/low temperature limits, chemical- and thermal-resistant top plate, and transparent tank for excellent visibility.