This item is now discontinued. We recommend the following as suggested replacements: WZ-12101-61

PolyScience LX Immersion Circulator; 120 VAC, 60 Hz

Representative image only.
PolyScience Mfr # LXC-9A11B – Item # WZ-12101-62
Designed for milder temperatures, smaller vessels, and lower flow rate applications
  • Large, user-friendly, backlit digital display
  • Clamp is adjustable for immersion depth and works on flat or curved-wall tanks
  • Heating coil and circulation pump are built into the housing
  • Built for laboratory and industrial use

SPECS & DESCRIPTION

Specifications & Description

  • Bath TypeImmersion Circulators
  • Reservoir Capacity (L)20
  • Max Temperature (° C)98
  • Min Temperature (° C)Ambient 10
  • Pump TypeForce
  • Low-Level CutoffYes
  • High-Temperature CutoffYes
  • Width (in)3 3/4
  • Length (in)14
  • Height (in)6 1/2
  • Display TypeLCD
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionImmersion Circulator; 120 VAC, 60 Hz
Two year warranty

Includes

6-ft (1.8-m) power cord with three-prong plug (European plug on 240 VAC unit).

Key Features

  • Large, user-friendly, backlit digital display
  • Clamp is adjustable for immersion depth and works on flat or curved-wall tanks
  • Heating coil and circulation pump are built into the housing
  • Built for laboratory and industrial use

More About this Item

Moderate temperature range is ideal for constant temperature, sample preparation, analytical methods, incubation, cell culture, enzyme/tissue studies, and fermentation applications. This streamlined circulator features a large, easy-to-read display that shows both actual and set point temperatures, as well as icons for heat-on and circulating-on. Built-in timer alerts you when samples are ready or processes are complete. Simple four-button control provides operation of on/off/standby, set point value, Fahrenheit/Celsius selection, timer setting, high/low temperature alarm set points, and offset calibration value.

Maximum temperature of 98°C works with both plastic and metal tanks, and the 6 L/min maximum flow rate is optimal for circulating water or low-viscosity fluids. Adjustable integrated mounting clamp fits on curved or flat walls up to 1 inch (2.5 cm) thick. Compact design is maintenance free and maximizes available reservior space. Protective housing shields pump and heater, yet offers easy access for cleaning — samples are safe from contacting the coil or clogging the pump.

REVIEWS

REVIEW SNAPSHOT®

by PowerReviews
PolyScience LX Immersion Circulator; 120 VAC, 60 Hz
 
1.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

(2 of 2 customers found this review helpful)

 
1.0

Unwanted Timer

By 

from Newark, DE

Verified Reviewer

Pros

  • price
  • size

Cons

  • Annoying Timer Cannot Be

Best Uses

    Comments about PolyScience LX Immersion Circulator; 120 VAC, 60 Hz:

    Annoying timer cannot be turned off. The advertising for this unit did not even mention a timer.

    • Was this a gift?:
    • No

    Displaying review 1

    Back to top

    Q&A

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    2 Questions | 2 Answers
    Displaying questions 1-2
    • Q:

      can this cool water to a temp of 19°C?
      Asked on 2/21/2017 by Gavin

      1 answer

      • STAFF EXPERT

        A:

        This is a heating immersion circulator with a range of Ambient + 10° to 98°C. It is not a chiller so it will not be able to lower the temperature to 19°C.

        Answered on 2/22/2017 by Polo from Technical Support
    • Q:

      Can this be placed on the edge of a 5-gallon pail/bucket?
      Asked on 1/8/2016 by Aimee

      1 answer

      • STAFF EXPERT

        A:

        This unit has a straight mounting clip to the ability to be mounted on the side of a tank. If the bucket or pail is round, the clamp may not fit properly on the lip. The bucket must also be taller than 6.5".

        Answered on 1/8/2016 by Katie from Technical Support
    Displaying questions 1-2

    Do you have a question about this product? 

    CUSTOMER SUPPORT

    Need Help with this product?

    Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

    Call Us At+44 (0)1480 272279
    Live chat