Plenum Coarse Bubble Diffuser, 304 SS, 1-ft L
Representative image only.
Item # WZ-70025-10
Increase oxygen into your water
  • Ideal for water and wastewater treatment
  • Provides oxygenation and mixing capabilities essential for biological treatment processes
PRODUCT OPTIONS
£122.19GBP / Each
£146.63GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Thread Size NPT (in)3/4
  • Max Flow Range (CFM)25
  • Min Flow Range (CFM)2
  • Material304 Stainless Steel
Key Features
  • Ideal for water and wastewater treatment
  • Provides oxygenation and mixing capabilities essential for biological treatment processes
More About this Item
Diffuser aeration systems provide increased oxygen content to wastewater in treatment facilities. This process creates an environment where microorganisms biologically decompose soluble organic contaminants in wastewater.
Diffusers are also used to diffuse digester gases such as hydrogen sulfide and sulphur dioxide; for the introduction of ozone for the purpose of oxidizing and destroying bacterial and organic chemical contaminants; and for carbon dioxide injection for recarbonization and pH adjustment. Order air blowers separately to complete your system.