This item is now discontinued. Please browse similar items

Platinum Cover for 25 mL Standard Form Crucibles, 1/ea

Representative image only.
Mfr # GGCS25LID – Item # WZ-17831-45
Prohibit contamination at high temperatures up to 2192°F (1200°C)
  • Constructed of durable platinum

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)25
  • Cap MaterialPlatinum
  • AutoclavableYes
  • DescriptionPlatinum Cover for 25 mL Standard Form Crucibles, 1/ea

Key Features

  • Constructed of durable platinum

More About this Item

Platinum offers oxidation resistance in air and oxygen; and corrosion resistance in acids and melted salts. Lids, crucibles, and tongs sold separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat