This item is now discontinued. Please browse similar items
Phenix Technologies PM15-2 15k VDC Hipot-Megohmeter, 120V AC
Representative image only.
Phenix Technologies Mfr # PM15-2 – Item # WZ-20023-55
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description15k VDC Hipot-Megohmeter, 120V AC
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • Integral megohmmeter measures insulation resistance up to 500,000 megohms at 2.5 KVDC or up to 1,000,000 megohms @ 5.0 KVDC
  • Voltage output continuously adjustable up to 5,000 VAC or 6,000 VDC
  • “Zero Start” voltage interlock
  • Large, easy-to-read analog meters
  • Leakage current measurements from 0.1 to 5,000 microamps DC and from 1.0 to 5.0 milliamps AC
  • Adjustable overcurrent trip point
  • Rotary switch selection simplifies operation
  • Lightweight, portable and enclosed in a rugged, fiberglass carrying case
  • Model PM6 has same specifications as PAD56, less AC output capability