This item is now discontinued. Please browse similar items
Phenix Technologies 640-0.4P Portable AC Dielectric Test Set, 40kVAC, Vacuum Bottle
Representative image only.
Phenix Technologies Mfr # 640-0.4P – Item # WZ-20023-57
SPECS & DESCRIPTION
Specifications & Description
  • AC Current Range (mA)0 to 10
  • Max Voltage (Voltage)40000
  • DescriptionPortable AC Dielectric Test Set, 40kVAC, Vacuum Bottle
More About this Item
  • Now offering a premier line of 40, 50, and 60 kV models, providing test voltages for a wide range of electrical equipment
  • Featuring 3 1/2 digit, digital LCD display output voltmeter and currentmeter for easy reading even in direct sunlight
  • High accuracy ±1% FS
  • Ruggedly constructed cases for years of field use
  • Lightweight portability
  • Zero start (safety interlock feature)
  • Output termination: toroid with output lead
  • Adjustable overcurrent trip from 10% to 110% of rating
  • Internal short circuit protection
PX299640
  • 40 kV AC, 10 mA hipot designed primarily for vacuum bottle testing; however, it can be used for testing breakers, switchgear, and many othe
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
1 Question | 1 Answer
Displaying question 1
  • Q:
    Will this meter record results. If so are they downloadable and is there software provided for that purpose?
    Asked on 6/22/2011 by Anonymous from San Diego CA
    1 answer
    • STAFF EXPERT
      A:
      This tester does not have data logging capabilities.
      Answered on 6/23/2011 by Chris from Technical Support
Displaying question 1
Do you have a question about this product? 