Peter Swan SW340 VF Freezer, Heavy-Duty Upright 340-liter, -20C, 230V
Representative image only.
Peter Swan Mfr # SW340 VF – Item # WZ-44444-54
Economical cold storage for your laboratory
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)20
  • Freezer Capacity (Liters)566
  • Min Temperature - Refrigerator (° F)32
  • Max Temperature - Refrigerator (° F)50
  • Min Temperature - Refrigerator (° C)0
  • Max Temperature - Refrigerator (° C)10
  • Number of Doors1
  • Number of Shelves5
  • Power (VAC)240
  • Power (Hz)50
  • Power (watts)480
  • DescriptionFreezer, Heavy-Duty Upright 340-liter, -20C, 230V
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
These general-purpose laboratory refrigerators and freezers are designed to meet most laboratory requirements, with refrigerators maintaining a temperature of 4degC and freezers maintaining a temperature of -20°C.
Research Laboratory refrigerators and freezers are available in either standard duty or heavy duty models. These models are internally spark-free and feature a standard door lock for added protection of samples. The heavy duty models are designed for busy laboratories, incorporating an upgraded refrigeration system with extra thick insulation, assisting in quicker recovery times when door is opened.
Electronic temperature control freezers also feature a standard door lock. Models 44444-60, 44444-62, and 44444-63 feature fixed freezing shelves and are internally spark-free.