This item is now discontinued. Please browse similar items

nVent Hoffman HB160816G040 Enclosure HB16 8000 BTU 115V 1 PH

Representative image only.
nVent Hoffman Mfr # HB160816G040 – Item # WZ-10500-71
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • MaterialSteel
  • Outside Width (in)17
  • Outside Height (in)15.25
  • Outside Depth (in)25.1875
  • DescriptionEnclosure HB16 8000 BTU 115V 1 PH
  • Warranty1 Year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Parts and accessories to keep your Pentair enclosure working reliably.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat