This item is now discontinued. Please browse similar items
nVent Hoffman A1412CHNFSS6 Stainless Steel Junction Box, Continuous Hinge, type 4X
Representative image only.
nVent Hoffman Mfr # A1412CHNFSS6 – Item # WZ-10502-70
Protects components from indoor or outdoor corrosive environments
  • Seams continuously welded and ground smooth
  • Seamless foam-in-place gasket
  • Features stainless steel screws and clamps
  • Bonding provision on door and body
  • Ratings: NEMA Type 4X, IP66
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Outside Width (in)12
  • Outside Height (in)14
  • Outside Depth (in)6
  • DescriptionStainless Steel Junction Box, Continuous Hinge, type 4X
  • Warranty1 Year
1 Year WarrantyCanadian Standards AssociationIP 66 Rated
Key Features
  • Seams continuously welded and ground smooth
  • Seamless foam-in-place gasket
  • Features stainless steel screws and clamps
  • Bonding provision on door and body
  • Ratings: NEMA Type 4X, IP66
  • 1 year warranty
More About this Item
Used in either indoor or outdoor applications, these enclosures combine a rugged continuous hinge, seamless foam-in-place gasket and stainless steel screw-down clamps for a reliable seal that protects components from corrosive environments. The cover and sides of the body have a smooth #4 brushed finish.